2-Methyl-4-ethylthiazoline ≥88.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAM0689-25MLEA 700 CNY
TCIAM0689-25ML
2-Methyl-4-ethylthiazoline ≥88.0% (通过GC)
2-Methyl-4-ethylthiazoline
分子式: C₆H₁₁NS
MDL编号: MFCD00015468
CAS号: 4293-61-2
EINECS: 224-297-3
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 88.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR