1-Methyl-4-piperidoneoxime hydrochloride ≥97.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAM0838-1GEA 540 CNY
TCIAM0838-1G
1-Methyl-4-piperidoneoxime hydrochloride ≥97.0% (通过滴定分析)
1-Methyl-4-piperidoneoxime hydrochloride
分子式: C₆H₁₂N₂O
MDL编号: MFCD00060190
CAS号: 84540-61-4
EINECS: 283-155-9

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR