(R)-(+)-2-(ɑ-Methylbenzylamino)-5-nitropyridine ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAM0909-5GEA 0 CNY
TCIAM0909-5G
(R)-(+)-2-(ɑ-Methylbenzylamino)-5-nitropyridine ≥98.0% (通过滴定分析)
(R)-(+)-2-(alpha-Methylbenzylamino)-5-nitropyridine
分子式: C₁₃H₁₃N₃O₂
分子量: 243,27 g/mol
MDL编号: MFCD00060067
CAS号: 64138-65-4

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 81.0 to 84.0 deg-C
Specific rotation [a]20/D +145.0 to +148.0 deg (C=2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR