(S)-(+)-5-Methyl-1-heptanol ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAM0965-1MLEA 0 CNY
TCIAM0965-1ML
(S)-(+)-5-Methyl-1-heptanol ≥97.0% (通过GC)
(S)-(+)-5-Methyl-1-heptanol
分子式: C₈H₁₈O
分子量: 130,23 g/mol
沸点: 84°C
MDL编号: MFCD00135168
CAS号: 57803-73-3

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR