4-Methyl-6,7-methylenedioxycoumarin

供应商: TCI EUROPE

TCIAM0971-1GEA 775 CNY
TCIAM0971-1G
4-Methyl-6,7-methylenedioxycoumarin
4-Methyl-6,7-methylenedioxycoumarin
分子式: C₁₁H₈O₄
分子量: 204,18 g/mol
MDL编号: MFCD00143343
CAS号: 15071-04-2

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 180.0 to 183.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR