4-Methoxy-4'-nitrostilbene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAM0999-5GEA 0 CNY
TCIAM0999-5G
4-Methoxy-4'-nitrostilbene ≥98.0% (通过GC)
4-Methoxy-4'-nitrostilbene
分子式: C₁₅H₁₃NO₃
MDL编号: MFCD00143335
CAS号: 1472-68-0

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 134.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR