ɑ-Methyllevulinic acid ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAM1058-5GEA
0
CNY
TCIAM1058-5G
ɑ-Methyllevulinic acid ≥98.0% (通过GC)
alpha-Methyllevulinic acid
分子式:
C₆H₁₀O₃ 分子量: 130,14 g/mol 沸点: 165°C |
MDL编号:
MFCD00143584 CAS号: 6641-83-4 |
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...