4-Methyl-2(5H)-furanone ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAM1079-5MLEA 4150 CNY
TCIAM1079-5ML
4-Methyl-2(5H)-furanone ≥97.0% (通过GC)
4-Methyl-2(5H)-furanone
分子式: C₅H₆O₂
沸点: 113°C
闪点: 95°C
MDL编号: MFCD00191546
CAS号: 6124-79-4

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR