3-Methyl-5-phenylpyridine ≥95.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAM1094-5GEA 0 CNY
TCIAM1094-5G
3-Methyl-5-phenylpyridine ≥95.0% (通过GC)
3-Methyl-5-phenylpyridine
分子式: C₁₂H₁₁N
分子量: 169,23 g/mol
沸点: 160°C
MDL编号: MFCD00191465
CAS号: 10477-94-8

现在订购

产品技术规格

Appearance Light yellow to Brown to Dark green clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR