4,5-Methylenedithio-1,3-dithiole-2-thione ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAM1112-1GEA 0 CNY
TCIAM1112-1G
4,5-Methylenedithio-1,3-dithiole-2-thione ≥98.0% (通过GC)
4,5-Methylenedithio-1,3-dithiole-2-thione
分子式: C₄H₂S₅
MDL编号: MFCD00137884
CAS号: 70800-59-8

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR