7-Methoxy-3-phenylsulphonyl-1(3H)-isobenzofuranone ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAM1130-1GEA 0 CNY
TCIAM1130-1G
7-Methoxy-3-phenylsulphonyl-1(3H)-isobenzofuranone ≥98.0% (通过HPLC)
7-Methoxy-3-phenylsulphonyl-1(3H)-isobenzofuranone
分子式: C₁₅H₁₂O₅S
分子量: 304,32 g/mol
MDL编号: MFCD00191458
CAS号: 65131-09-1

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 186.0 to 190.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR