(±)-α-甲氧基-α-(三氟甲基)苯乙腈 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAM1163-5GEA 0 CNY
TCIAM1163-5G
(±)-α-甲氧基-α-(三氟甲基)苯乙腈 ≥98.0% (通过GC)
(±)-α-甲氧基-α-(三氟甲基)苯乙腈
分子式: C₁₀H₈F₃NO
分子量: 215,18 g/mol
闪点: 67°C
MDL编号: MFCD00001844
CAS号: 80866-87-1
EINECS: 279-591-4

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR