3-(Methoxycarbonyl)-2H-cyclohepta[b]furan-2-one ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAM1188-1GEA
0
CNY
TCIAM1188-1G
3-(Methoxycarbonyl)-2H-cyclohepta[b]furan-2-one ≥98.0% (通过GC)
3-(Methoxycarbonyl)-2H-cyclohepta[b]furan-2-one
分子式:
C₁₁H₈O₄ 分子量: 204,18 g/mol |
MDL编号:
MFCD00671552 CAS号: 50603-71-9 |
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...