3-Mercaptoisobutyric acid ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAM1200-25GEA 0 CNY
TCIAM1200-25G
3-Mercaptoisobutyric acid ≥98.0% (通过GC,滴定分析)
3-Mercaptoisobutyric acid
分子式: C₄H₈O₂S
分子量: 120,17 g/mol
沸点: 122°C
MDL编号: MFCD00671555
CAS号: 26473-47-2
EINECS: 247-726-6

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR