2-Methyl-1,2'-dinaphthyl ketone

供应商: TCI EUROPE

TCIAM1284-1GEA 0 CNY
TCIAM1284-1G
2-Methyl-1,2'-dinaphthyl ketone
2-Methyl-1,2'-dinaphthyl ketone
分子式: C₂₂H₁₆O
分子量: 296,37 g/mol
MDL编号: MFCD01321179
CAS号: 110876-52-3

现在订购

产品技术规格

Appearance White to Yellow to Green powder to crystaline
Purity (GC) min. 98.0 %
Melting point 139.0 to 142.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR