2-Methylnaphth[1,2-d]oxazole ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAM1290-10GEA
0
CNY
TCIAM1290-10G
2-Methylnaphth[1,2-d]oxazole ≥98.0% (通过GC)
2-Methylnaphth[1,2-d]oxazole
分子式:
C₁₂H₉NO 分子量: 183,21 g/mol 沸点: 300°C |
MDL编号:
MFCD00051328 CAS号: 85-15-4 EINECS: 201-589-9 |
产品技术规格
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...