5-Mercapto-1-(4-methoxyphenyl)-1H-tetrazole ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Danger
|
TCIAM1292-10GEA
0
CNY
TCIAM1292-10G
5-Mercapto-1-(4-methoxyphenyl)-1H-tetrazole ≥98.0% (通过滴定分析)
5-Mercapto-1-(4-methoxyphenyl)-1H-tetrazole
分子式:
C₈H₈N₄OS 分子量: 208,24 g/mol |
MDL编号:
MFCD01321180 CAS号: 13980-76-2 UN号: 1325 ADR: 4.1,II |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...