β-Methylcholine chloride ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAM1293-25GEA 0 CNY
TCIAM1293-25G
β-Methylcholine chloride ≥98.0% (通过滴定分析)
beta-Methylcholine chloride
分子式: C₆H₁₆ClNO
分子量: 153,65 g/mol
MDL编号: MFCD01669408
CAS号: 2382-43-6
EINECS: 219-183-5

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR