4-甲氧基-3,5-二甲基-2-羟甲基吡啶 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAM1327-5GEA
0
CNY
TCIAM1327-5G
4-甲氧基-3,5-二甲基-2-羟甲基吡啶 ≥98.0% (通过GC)
4-甲氧基-3,5-二甲基-2-羟甲基吡啶
分子式:
C9H13NO2 分子量: 167,2100 g/mol |
MDL编号:
MFCD01076195 CAS号: 86604-78-6 EINECS: 289-258-5 |
产品技术规格
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 63.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...