(1S)-(-)-10-Mercaptoborneol ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAM1341-100MGEA 0 CNY
TCIAM1341-100MG
(1S)-(-)-10-Mercaptoborneol ≥97.0% (通过GC)
(1S)-(-)-10-Mercaptoborneol
分子式: C₁₀H₁₈OS
分子量: 186,31 g/mol
MDL编号: MFCD02093482
CAS号: 71242-59-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 83.0 to 87.0 deg-C
Specific rotation [a]20/D -5.0 to -7.0 deg (C=2, CHCl2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR