2-甲巯基吩噻嗪 ≥98.0% (通过HPLC)

供应商: TCI EUROPE
Warning

Synonyms: 2-Methylsulfanyl-10H-phenothiazine

TCIAM1387-25GEA 0 CNY
TCIAM1387-25G
2-甲巯基吩噻嗪 ≥98.0% (通过HPLC)
2-甲巯基吩噻嗪
分子式: C₁₃H₁₁NS₂
分子量: 245,36 g/mol
MDL编号: MFCD00799974
CAS号: 7643-08-5
EINECS: 231-581-0

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 140.0 to 143.0 deg-C
Solubility in Acetone very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR