2,2'-Methylenebisbenzothiazole ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAM1390-5GEA 445 CNY
TCIAM1390-5G
2,2'-Methylenebisbenzothiazole ≥97.0% (通过GC)
2,2'-Methylenebisbenzothiazole
分子式: C₁₅H₁₀N₂S₂
MDL编号: MFCD00051395
CAS号: 1945-78-4
EINECS: 217-744-9

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR