2-(三甲基硅氧基)乙基甲基丙烯酸盐 ≥96.0% (通过GC) stabilised
供应商: TCI EUROPE
|
Warning
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
TCIAM1391-25GEA
0
CNY
TCIAM1391-25G
2-(三甲基硅氧基)乙基甲基丙烯酸盐 ≥96.0% (通过GC) stabilised
2-(三甲基硅氧基)乙基甲基丙烯酸盐
分子式:
C₉H₁₈O₃Si 分子量: 202,33 g/mol 沸点: 72°C 闪点: 76°C |
MDL编号:
MFCD00053869 CAS号: 17407-09-9 EINECS: 241-432-1 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...