1-(Methoxymethoxy)naphthalene ≥97.0% (通过GC)
供应商: TCI EUROPE
TCIAM1454-5GEA
0
CNY
TCIAM1454-5G
1-(Methoxymethoxy)naphthalene ≥97.0% (通过GC)
1-(Methoxymethoxy)naphthalene
分子式:
C₁₂H₁₂O₂ 分子量: 188,23 g/mol 沸点: 296°C |
MDL编号:
MFCD06797119 CAS号: 7382-37-8 |
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...