2-(N-Methyl-2-pyridylamino)ethanol ≥98.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAM1468-5GEA
0
CNY
TCIAM1468-5G
2-(N-Methyl-2-pyridylamino)ethanol ≥98.0% (通过GC,滴定分析)
2-(N-Methyl-2-pyridylamino)ethanol
分子式:
C₈H₁₂N₂O 分子量: 152,20 g/mol |
MDL编号:
MFCD02667633 CAS号: 122321-04-4 |
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...