4-Methoxyphenyl-3-O-allyl-β-D-galactopyranoside ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAM1482-5GEA 790 CNY
TCIAM1482-5G
4-Methoxyphenyl-3-O-allyl-β-D-galactopyranoside ≥98.0% (通过HPLC)
4-Methoxyphenyl-3-O-allyl-beta-D-galactopyranoside
分子式: C₁₆H₂₂O₇
存储条件: Freezer
MDL编号: MFCD06797127
CAS号: 144985-19-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 141.0 to 145.0 deg-C
Specific rotation [a]20/D -8.0 to -11.0 deg (C=1,methanol)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR