4,4′-亚甲基双(苯磺酰氯) ≥95.0% (通过滴定分析)

供应商: TCI EUROPE
Danger

Synonyms: 二苯基甲烷-4,4′-二磺酰氯

TCIAM1556-1GEA 0 CNY
TCIAM1556-1G
4,4′-亚甲基双(苯磺酰氯) ≥95.0% (通过滴定分析)
4,4′-亚甲基双(苯磺酰氯)
分子式: C₁₃H₁₀Cl₂O₄S₂
分子量: 365,24 g/mol
CAS号: 3119-64-0
UN号: 3261
ADR: 8,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 95.0 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR