4-Methoxyphenyl-2-azido-3,6-di-O-benzyl-2-deoxy-β-D-glucopyranoside
供应商: TCI EUROPE
TCIAM1617-1GEA
1275
CNY
TCIAM1617-1G
4-Methoxyphenyl-2-azido-3,6-di-O-benzyl-2-deoxy-β-D-glucopyranoside
4-Methoxyphenyl-2-azido-3,6-di-O-benzyl-2-deoxy-beta-D-glucopyranoside
分子式:
C₂₇H₂₉N₃O₆ 存储条件: Freezer |
CAS号:
1272755-25-5 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Elemental analysis (Nitrogen) | 8.35 to 8.65 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...