4'-甲氧基甲酰苯胺 ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAM1623-5GEA 505 CNY
TCIAM1623-5G
4'-甲氧基甲酰苯胺 ≥98.0% (通过GC)
4'-甲氧基甲酰苯胺
分子式: C₈H₉NO₂
分子量: 151,17 g/mol
沸点: 159°C
MDL编号: MFCD00086774
CAS号: 5470-34-8
EINECS: 226-802-2

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR