4-Methoxyphenyl-3-O-benzyl-β-D-glucopyranoside ≥98.0% (通过HPLC)
供应商: TCI EUROPE
TCIAM1641-1GEA
0
CNY
TCIAM1641-1G
4-Methoxyphenyl-3-O-benzyl-β-D-glucopyranoside ≥98.0% (通过HPLC)
4-Methoxyphenyl-3-O-benzyl-beta-D-glucopyranoside
分子式:
C₂₀H₂₄O₇ 分子量: 376,41 g/mol 存储条件: Freezer |
CAS号:
303127-80-2 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 141.0 to 146.0 deg-C |
Specific rotation [a]20/D | -50.0 to -55.0 deg (C=0.1chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...