3-亚甲基-2-降冰片酮 ≥97.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAM1656-5GEA
0
CNY
TCIAM1656-5G
3-亚甲基-2-降冰片酮 ≥97.0% (通过GC)
3-亚甲基-2-降冰片酮
分子式:
C₈H₁₀O 分子量: 122,17 g/mol 沸点: 70°C 闪点: 55°C |
MDL编号:
MFCD00151643 CAS号: 5597-27-3 EINECS: 227-009-4 UN号: 1224 ADR: 3,III |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...