7-甲氧基-1,4-苯并噻嗪-3(4H)-酮 ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAM1662-5GEA 0 CNY
TCIAM1662-5G
7-甲氧基-1,4-苯并噻嗪-3(4H)-酮 ≥98.0% (通过GC)
7-甲氧基-1,4-苯并噻嗪-3(4H)-酮
分子式: C₉H₉NO₂S
分子量: 195,24 g/mol
MDL编号: MFCD02660686
CAS号: 22726-30-3

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 183.0 to 187.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR