1-Methylpyrrolidine-2-thione ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAM1748-5GEA 0 CNY
TCIAM1748-5G
1-Methylpyrrolidine-2-thione ≥97.0% (通过GC)
1-Methylpyrrolidine-2-thione
分子式: C₅H₉NS
分子量: 115,19 g/mol
沸点: 145°C
溶点: 16°C
MDL编号: MFCD00966332
CAS号: 10441-57-3

现在订购

产品技术规格

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR