3-Methyl-7-propylxanthine ≥96.0% (以总氮计)
供应商: TCI EUROPE
TCIAM1756-25GEA
0
CNY
TCIAM1756-25G
3-Methyl-7-propylxanthine ≥96.0% (以总氮计)
3-Methyl-7-propylxanthine
分子式:
C9H12N4O2 分子量: 208,2172 g/mol |
MDL编号:
MFCD02089337 CAS号: 55242-64-3 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...