2-甲基-3-呋喃硫醇 ≥95.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAM1847-1GEA 0 CNY
TCIAM1847-1G
2-甲基-3-呋喃硫醇 ≥95.0% (通过GC)
2-甲基-3-呋喃硫醇
分子式: C₅H₆OS
分子量: 114,16 g/mol
闪点: 37°C
存储条件: Refrigerator
MDL编号: MFCD00010280
CAS号: 28588-74-1
EINECS: 249-094-7
UN号: 1228
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR