10-Methoxy-5H-dibenzo[b,f]azepine ≥98.0% (通过HPLC,总氮)
供应商: TCI EUROPE
TCIAM1895-25GEA
0
CNY
TCIAM1895-25G
10-Methoxy-5H-dibenzo[b,f]azepine ≥98.0% (通过HPLC,总氮)
10-Methoxy-5H-dibenzo[b,f]azepine
分子式:
C₁₅H₁₃NO 分子量: 223,28 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00101079 CAS号: 4698-11-7 EINECS: 225-172-6 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...