5-甲基胞苷 ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAM1931-1GEA 900 CNY
TCIAM1931-1G
5-甲基胞苷 ≥98.0% (通过HPLC,滴定分析)
5-甲基胞苷
分子式: C₁₀H₁₅N₃O₅
分子量: 257,25 g/mol
溶点: 211°C
MDL编号: MFCD00006548
CAS号: 2140-61-6
EINECS: 218-390-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +13.0 to +17.0 deg (C=2, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR