1-氯甲酰基-3-甲磺酰基-2-咪唑烷酮 ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Danger

TCIAM1944-25GEA 0 CNY
TCIAM1944-25G
1-氯甲酰基-3-甲磺酰基-2-咪唑烷酮 ≥98.0% (通过滴定分析)
1-氯甲酰基-3-甲磺酰基-2-咪唑烷酮
分子式: C₅H₇ClN₂O₄S
分子量: 226,63 g/mol
MDL编号: MFCD00272360
CAS号: 41762-76-9
EINECS: 255-540-1
UN号: 3261
ADR: 8,II

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR