3-Methyl-3-(2-oxopropyl)-2,4,7-trioxabicyclo[3.3.0]octane ≥95.0% (通过GC)
供应商: TCI EUROPE
TCIAM1978-5GEA
0
CNY
TCIAM1978-5G
3-Methyl-3-(2-oxopropyl)-2,4,7-trioxabicyclo[3.3.0]octane ≥95.0% (通过GC)
3-Methyl-3-(2-oxopropyl)-2,4,7-trioxabicyclo[3.3.0]octane
分子式:
C₉H₁₄O₄ 分子量: 186,21 g/mol |
CAS号:
1378825-86-5 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...