3-Mercaptopropyl silica gel 0,5 - 0,8 mmol/g
供应商: TCI EUROPE
Warning
TCIAM1979-25GEA 1125 CNY
TCIAM1979-25G
3-Mercaptopropyl silica gel 0,5 - 0,8 mmol/g
3-Mercaptopropyl silica gel
现在订购
产品技术规格
Appearance White to Almost white powder to crystal
Loading 0.5 to 0.8 mmol/g