5-甲基-2-苯基-2-己烯醛 ≥90.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAM1997-25GEA 0 CNY
TCIAM1997-25G
5-甲基-2-苯基-2-己烯醛 ≥90.0% (通过GC)
5-甲基-2-苯基-2-己烯醛
分子式: C₁₃H₁₆O
分子量: 188,27 g/mol
沸点: 100°C
MDL编号: MFCD00036615
CAS号: 21834-92-4
EINECS: 244-602-3

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR