2-(4-Methoxybenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAM2071-5GEA 0 CNY
TCIAM2071-5G
2-(4-Methoxybenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (通过GC,滴定分析)
2-(4-Methoxybenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
分子式: C₁₄H₂₁BO₃
分子量: 248,13 g/mol
存储条件: Refrigerator
MDL编号: MFCD10698526
CAS号: 475250-52-3

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR