9-Mesityl-2,7,10-trimethylacridinium perchlorate ≥98.0% (通过HPLC)

供应商: TCI EUROPE
Warning

TCIAM2072-1GEA 2225 CNY
TCIAM2072-1G
9-Mesityl-2,7,10-trimethylacridinium perchlorate ≥98.0% (通过HPLC)
9-Mesityl-2,7,10-trimethylacridinium perchlorate
分子式: C₂₅H₂₆ClNO₄
分子量: 439,94 g/mol
存储条件: Refrigerator
CAS号: 1216909-33-9
UN号: 1479
ADR: 5.1,III

现在订购

产品技术规格

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR