1-Methyl-1H-pyrazole-5-carbaldehyde ≥95.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAM2179-1GEA
0
CNY
TCIAM2179-1G
1-Methyl-1H-pyrazole-5-carbaldehyde ≥95.0% (通过GC)
1-Methyl-1H-pyrazole-5-carbaldehyde
分子式:
C₅H₆N₂O 分子量: 110,1100 g/mol 沸点: 53°C |
MDL编号:
MFCD03419801 CAS号: 27258-33-9 |
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...