5-Methyl-N-carbobenzoxy-L-glutamate ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAM2269-5GEA 0 CNY
TCIAM2269-5G
5-Methyl-N-carbobenzoxy-L-glutamate ≥98.0% (通过HPLC,滴定分析)
5-Methyl-N-carbobenzoxy-L-glutamate
分子式: C₁₄H₁₇NO₆
分子量: 295,29 g/mol
存储条件: Refrigerator
MDL编号: MFCD00191090
CAS号: 4652-65-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 68.0 to 72.0 deg-C
Specific rotation [a]20/D -11.0 to -13.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR