4-Nitrobenzoylglycylglycine ≥99.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAN0177-1GEA 0 CNY
TCIAN0177-1G TCIAN0177-25G
4-Nitrobenzoylglycylglycine ≥99.0% (通过滴定分析)
4-Nitrobenzoylglycylglycine
分子式: C₁₁H₁₁N₃O₆
分子量: 281,22 g/mol
溶点: 218°C
MDL编号: MFCD00059183
CAS号: 78196-53-9

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR