3'-Nitro-4-dimethylaminoazobenzene ≥97.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAN0191-25GEA 0 CNY
TCIAN0191-25G
3'-Nitro-4-dimethylaminoazobenzene ≥97.0% (通过滴定分析)
3'-Nitro-4-dimethylaminoazobenzene
分子式: C₁₄H₁₄N₄O₂
分子量: 270,29 g/mol
MDL编号: MFCD00059825
CAS号: 3837-55-6

现在订购

产品技术规格

Appearance Orange to Red powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 159.0 to 163.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR