新胭脂红 ≥85.0% (通过UV/Vis)

供应商: TCI EUROPE

Synonyms: 酸性红 18丽春红 4R

TCIAN0309-25GEA 0 CNY
TCIAN0309-25G
新胭脂红 ≥85.0% (通过UV/Vis)
新胭脂红
分子式: C₂₀H₁₄N₂O₁₀S₃
分子量: 604,46 g/mol
MDL编号: MFCD00004084
CAS号: 2611-82-7
EINECS: 220-036-2

现在订购

产品技术规格

Appearance Dark red powder to crystal
Purity (UV-vis method) min. 85.0 %
Lambda max. 506.0 to 510.0 nm (0.02mol/L AcONH4 sol.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR