2-Propoxynaphthalene

供应商: TCI EUROPE

TCIAN0432-25GEA 0 CNY
TCIAN0432-25G
2-Propoxynaphthalene
2-Propoxynaphthalene
分子式: C₁₃H₁₄O
分子量: 186,25 g/mol
沸点: 305°C
存储条件: Refrigerator
MDL编号: MFCD00021610
CAS号: 19718-45-7

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR