2-Nitrophenetole ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAN0489-25GEA 0 CNY
TCIAN0489-25G
2-Nitrophenetole ≥98.0% (通过GC)
2-Nitrophenetole
分子式: C₈H₉NO₃
分子量: 167,16 g/mol
沸点: 275°C
MDL编号: MFCD00053603
CAS号: 610-67-3
EINECS: 210-232-6

现在订购

产品技术规格

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR